3-Chlor-5-(pentafluorthio)anilin 98%

Lieferant: Apollo Scientific

PC302834-1G PC302834-5G PC302834-10G
APOSPC302834-1GEA 422 EUR
APOSPC302834-1G APOSPC302834-5G APOSPC302834-10G
3-Chlor-5-(pentafluorthio)anilin 98%
3-Chlor-5-(pentafluorthio)anilin
Formel: C₆H₅ClF₅NS
Molecular Weight: 253,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16652453
CAS-Nummer: 1240257-97-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR