3-Ethoxy-2,4-difluorbenzylalkohol 97%
Lieferant: Apollo Scientific
PC302838-10G
PC302838-5G
PC302838-1G
APOSPC302838-10GEA
799
EUR
APOSPC302838-10G
APOSPC302838-5G
APOSPC302838-1G
3-Ethoxy-2,4-difluorbenzylalkohol 97%
3-Ethoxy-2,4-difluorbenzylalkohol
Formel:
C₉H₁₀F₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258728 CAS-Nummer: 1017778-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...