3-Fluor-2-methoxybenzamid 97%
Lieferant: Apollo Scientific
PC302840-1G
PC302840-5G
APOSPC302840-1GEA
132
EUR
APOSPC302840-1G
APOSPC302840-5G
3-Fluor-2-methoxybenzamid 97%
3-Fluor-2-methoxybenzamid
Formel:
C₈H₈FNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06637313 CAS-Nummer: 874830-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...