3-Fluor-5-(trifluormethoxy)anilin

Lieferant: Apollo Scientific
Achtung

PC302847-1G PC302847-5G PC302847-250MG
APOSPC302847-1GEA 410 EUR
APOSPC302847-1G APOSPC302847-5G APOSPC302847-250MG
3-Fluor-5-(trifluormethoxy)anilin
3-Fluor-5-(trifluormethoxy)anilin
Formel: C₇H₅F₄NO
Lagertemperatur: Raumtemperatur
MDL: MFCD18783229
CAS-Nummer: 1352999-96-2
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR