3-Methoxy-4-(trifluormethyl)benzonitril 98%

Lieferant: Apollo Scientific
Achtung

PC302851-10G PC302851-5G PC302851-1G
APOSPC302851-10GEA 405 EUR
APOSPC302851-10G APOSPC302851-5G APOSPC302851-1G
3-Methoxy-4-(trifluormethyl)benzonitril 98%
3-Methoxy-4-(trifluormethyl)benzonitril
Formel: C₉H₆F₃NO
Molecular Weight: 201,15 g/mol
Schmelzpunkt: 52…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13185685
CAS-Nummer: 447-93-8
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR