4-Brom-3-fluor-2-(trifluormethyl)anilin 97%

Lieferant: Apollo Scientific

PC302856-1G PC302856-5G
APOSPC302856-1GEA 183 EUR
APOSPC302856-1G APOSPC302856-5G
4-Brom-3-fluor-2-(trifluormethyl)anilin 97%
4-Brom-3-fluor-2-(trifluormethyl)anilin
Formel: C₇H₄BrF₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD11519308
CAS-Nummer: 1092460-59-7
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR