4-Chloro-2-trifluoromethoxy-DL-phenylalanine

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-trifluoromethoxy-DL-phenylalanine

PC302857-1G PC302857-5G
APOSPC302857-1GEA 159 EUR
APOSPC302857-1G APOSPC302857-5G
4-Chloro-2-trifluoromethoxy-DL-phenylalanine
4-Chloro-2-trifluoromethoxy-DL-phenylalanine
Formel: C₁₀H₉ClF₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD19687214
CAS-Nummer: 1435806-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR