4-Chloro-2-(trifluoromethoxy)phenylacetonitrile 97%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-(trifluoromethoxy)phenylacetonitrile

PC302858-1G PC302858-5G
APOSPC302858-1GEA 159 EUR
APOSPC302858-1G APOSPC302858-5G
4-Chloro-2-(trifluoromethoxy)phenylacetonitrile 97%
4-Chloro-2-(trifluoromethoxy)phenylacetonitrile
Formel: C₉H₅ClF₃NO
Molecular Weight: 235,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18393878
CAS-Nummer: 1261861-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR