4-Chloro-2-fluoro-3-methoxyphenylacetonitrile 98%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-fluoro-3-methoxyphenylacetonitrile

PC302863-1G PC302863-5G
APOSPC302863-1GEA 159 EUR
APOSPC302863-1G APOSPC302863-5G
4-Chloro-2-fluoro-3-methoxyphenylacetonitrile 98%
4-Chloro-2-fluoro-3-methoxyphenylacetonitrile
Formel: C₉H₇ClFNO
Molecular Weight: 199,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687183
CAS-Nummer: 1323966-29-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR