5-Fluor-2-methoxybenzamid 98%
Lieferant: Apollo Scientific
PC302892-10G
PC302892-5G
PC302892-1G
APOSPC302892-10GEA
282
EUR
APOSPC302892-10G
APOSPC302892-5G
APOSPC302892-1G
5-Fluor-2-methoxybenzamid 98%
5-Fluor-2-methoxybenzamid
Formel:
C₈H₈FNO₂ Molecular Weight: 169,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06409124 CAS-Nummer: 704-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...