2-Chlor-4,6-bis(trifluormethyl)benzoesäure 98%

Lieferant: Apollo Scientific
Achtung

PC302911-1G PC302911-5G PC302911-10G
APOSPC302911-1GEA 180 EUR
APOSPC302911-1G APOSPC302911-5G APOSPC302911-10G
2-Chlor-4,6-bis(trifluormethyl)benzoesäure 98%
2-Chlor-4,6-bis(trifluormethyl)benzoesäure
Formel: C₉H₃ClF₆O₂
Molecular Weight: 292.56 g/mol
Schmelzpunkt: 148…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09025365
CAS-Nummer: 916420-47-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR