2-Ethoxy-3,5-difluorbenzylalkohol 98%

Lieferant: Apollo Scientific

PC302912-10G PC302912-5G PC302912-1G
APOSPC302912-10GEA 360 EUR
APOSPC302912-10G APOSPC302912-5G APOSPC302912-1G
2-Ethoxy-3,5-difluorbenzylalkohol 98%
2-Ethoxy-3,5-difluorbenzylalkohol
Formel: C₉H₁₀F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09258717
CAS-Nummer: 1017779-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR