2-Fluoro-4,6-bis(trifluoromethyl)benzoyl chloride

Lieferant: Apollo Scientific
Gefahr

PC302918-10G PC302918-5G
APOSPC302918-10GEA 328 EUR
APOSPC302918-10G APOSPC302918-5G
2-Fluoro-4,6-bis(trifluoromethyl)benzoyl chloride
2-Fluoro-4,6-bis(trifluoromethyl)benzoyl chloride
Formel: C₉H₂ClF₇O
Molecular Weight: 294,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258670
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR