2-Fluor-5-methylbenzylbromid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC302923-10G
PC302923-5G
PC302923-1G
APOSPC302923-10GEA
405
EUR
APOSPC302923-10G
APOSPC302923-5G
APOSPC302923-1G
2-Fluor-5-methylbenzylbromid 97%
2-Fluor-5-methylbenzylbromid
Formel:
C₈H₈BrF Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660227 CAS-Nummer: 886500-09-0 UN: 1759 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...