2-Methyl-5-(trifluoromethoxy)benzyl alcohol 98%

Lieferant: Apollo Scientific

Synonyme: 2-Methyl-5-(trifluoromethoxy)benzyl alcohol

PC302933-10G PC302933-5G PC302933-1G
APOSPC302933-10GEA 406 EUR
APOSPC302933-10G APOSPC302933-5G APOSPC302933-1G
2-Methyl-5-(trifluoromethoxy)benzyl alcohol 98%
2-Methyl-5-(trifluoromethoxy)benzyl alcohol
Formel: C₉H₉F₃O₂
Molecular Weight: 206,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18399766
CAS-Nummer: 1261478-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR