3-Chlor-5-fluor-4-methoxybenzonitril 98%

Lieferant: Apollo Scientific

PC302949-10G PC302949-5G PC302949-1G
APOSPC302949-10GEA 405 EUR
APOSPC302949-10G APOSPC302949-5G APOSPC302949-1G
3-Chlor-5-fluor-4-methoxybenzonitril 98%
3-Chlor-5-fluor-4-methoxybenzonitril
Formel: C₈H₅ClFNO
Molecular Weight: 185,59 g/mol
Schmelzpunkt: 71…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115866
CAS-Nummer: 886497-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR