3-Chloro-5-fluoro-4-methoxybenzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
PC302950-10G
PC302950-5G
PC302950-1G
APOSPC302950-10GEA
405
EUR
APOSPC302950-10G
APOSPC302950-5G
APOSPC302950-1G
3-Chloro-5-fluoro-4-methoxybenzyl bromide
3-Chloro-5-fluoro-4-methoxybenzyl bromide
Formel:
C₈H₇BrClFO Molecular Weight: 253.50 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115868 CAS-Nummer: 886497-36-5 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...