3'-Fluor-2'-methylacetophenon 98%

Lieferant: Apollo Scientific

PC302955-10G PC302955-5G PC302955-1G
APOSPC302955-10GEA 328 EUR
APOSPC302955-10G APOSPC302955-5G APOSPC302955-1G
3'-Fluor-2'-methylacetophenon 98%
3'-Fluor-2'-methylacetophenon
Formel: C₉H₉FO
Molecular Weight: 152.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832257
CAS-Nummer: 177942-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR