3-Methyl-5-(trifluormethoxy)benzamid 97%
Lieferant: Apollo Scientific
PC302963-10G
PC302963-5G
PC302963-1G
APOSPC302963-10GEA
449
EUR
APOSPC302963-10G
APOSPC302963-5G
APOSPC302963-1G
3-Methyl-5-(trifluormethoxy)benzamid 97%
3-Methyl-5-(trifluormethoxy)benzamid
Formel:
C₉H₈F₃NO₂ Molecular Weight: 219.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09025372 CAS-Nummer: 916420-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...