4'-(Pentafluorthio)acetophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC302967-5G
PC302967-1G
PC302967-25G
APOSPC302967-5GEA
986
EUR
APOSPC302967-5G
APOSPC302967-1G
APOSPC302967-25G
4'-(Pentafluorthio)acetophenon 98%
4'-(Pentafluorthio)acetophenon
Formel:
C₈H₇F₅OS Molecular Weight: 246,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16652408 CAS-Nummer: 401892-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...