4-Chloro-2-fluoro-3-methoxyphenylacetic acid 97%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-fluoro-3-methoxyphenylacetic acid

PC302975-1G PC302975-5G
APOSPC302975-1GEA 180 EUR
APOSPC302975-1G APOSPC302975-5G
4-Chloro-2-fluoro-3-methoxyphenylacetic acid 97%
4-Chloro-2-fluoro-3-methoxyphenylacetic acid
Formel: C₉H₈ClFO₃
Molecular Weight: 218,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687184
CAS-Nummer: 1323955-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR