4-Chloro-3-ethoxy-2-fluorobenzoic acid 98%
Lieferant: Apollo Scientific
PC302985-10G
PC302985-5G
PC302985-1G
APOSPC302985-10GEA
328
EUR
APOSPC302985-10G
APOSPC302985-5G
APOSPC302985-1G
4-Chloro-3-ethoxy-2-fluorobenzoic acid 98%
4-Chloro-3-ethoxy-2-fluorobenzoic acid
Formel:
C₉H₈ClFO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD19687215 CAS-Nummer: 1323966-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...