4-Ethoxy-2-fluorbenzonitril 98%
Lieferant: Apollo Scientific
PC302987-10G
PC302987-5G
PC302987-1G
APOSPC302987-10GEA
328
EUR
APOSPC302987-10G
APOSPC302987-5G
APOSPC302987-1G
4-Ethoxy-2-fluorbenzonitril 98%
4-Ethoxy-2-fluorbenzonitril
Formel:
C₉H₈FNO Molecular Weight: 165.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13196591 CAS-Nummer: 186191-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...