4-Fluor-3-(trifluormethoxy)phenylessigsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC302992-1G
PC302992-5G
APOSPC302992-1GEA
420
EUR
APOSPC302992-1G
APOSPC302992-5G
4-Fluor-3-(trifluormethoxy)phenylessigsäure 97%
4-Fluor-3-(trifluormethoxy)phenylessigsäure
Formel:
C₉H₆F₄O₃ Molecular Weight: 238,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660258 CAS-Nummer: 886501-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...