6'-Chlor-2',3'-difluoracetophenon 98%

Lieferant: Apollo Scientific
Achtung

PC303008-10G PC303008-5G PC303008-1G
APOSPC303008-10GEA 261 EUR
APOSPC303008-10G APOSPC303008-5G APOSPC303008-1G
6'-Chlor-2',3'-difluoracetophenon 98%
6'-Chlor-2',3'-difluoracetophenon
Formel: C₈H₅ClF₂O
Molecular Weight: 190.57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201051
CAS-Nummer: 1373920-86-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR