2-Chlor-3-(trifluormethyl)benzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-3-cyanobenzotrifluoride
PC303027-10G
PC303027-5G
PC303027-1G
PC303027-25G
APOSPC303027-10GEA
351
EUR
APOSPC303027-10G
APOSPC303027-5G
APOSPC303027-1G
APOSPC303027-25G
2-Chlor-3-(trifluormethyl)benzonitril 98%
2-Chlor-3-(trifluormethyl)benzonitril
Formel:
C₈H₃ClF₃N Molecular Weight: 205.57 Schmelzpunkt: 38…41 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115989 CAS-Nummer: 62584-32-1 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...