2-Chlor-6-fluor-3-methoxyanilin 98%

Lieferant: Apollo Scientific
Achtung

PC303030-10G PC303030-5G PC303030-1G
APOSPC303030-10GEA 608 EUR
APOSPC303030-10G APOSPC303030-5G APOSPC303030-1G
2-Chlor-6-fluor-3-methoxyanilin 98%
2-Chlor-6-fluor-3-methoxyanilin
Formel: C₇H₇ClFNO
Molecular Weight: 175,59 g/mol
Schmelzpunkt: 37…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09832265
CAS-Nummer: 1017777-58-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR