2-Methoxy-3-(trifluormethyl)benzylalkohol 98%
Lieferant: Apollo Scientific
PC303037-10G
PC303037-5G
APOSPC303037-10GEA
372
EUR
APOSPC303037-10G
APOSPC303037-5G
2-Methoxy-3-(trifluormethyl)benzylalkohol 98%
2-Methoxy-3-(trifluormethyl)benzylalkohol
Formel:
C₉H₉F₃O₂ Molecular Weight: 206,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832314 CAS-Nummer: 1017778-74-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...