2-Methoxy-5-(pentafluorothio)phenylacetonitrile 98%

Lieferant: Apollo Scientific

PC303040-1G PC303040-5G
APOSPC303040-1GEA 348 EUR
APOSPC303040-1G APOSPC303040-5G
2-Methoxy-5-(pentafluorothio)phenylacetonitrile 98%
2-Methoxy-5-(pentafluorothio)phenylacetonitrile
Formel: C₉H₈F₅NOS
Molecular Weight: 273,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16652489
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR