2-Methyl-5-(trifluoromethoxy)aniline 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-methyl-5-trifluoromethoxy-phenylamine, 2-methyl-5-(trifluoromethyloxy)aniline, 2-Methyl-5-trifluoromethoxyaniline

PC303047-5G PC303047-1G
APOSPC303047-5GEA 266 EUR
APOSPC303047-5G APOSPC303047-1G
2-Methyl-5-(trifluoromethoxy)aniline 98%
2-Methyl-5-(trifluoromethoxy)aniline
Formel: C₈H₈F₃NO
Molecular Weight: 191.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07777196
CAS-Nummer: 933674-93-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR