3-Chlor-4-(trifluormethyl)benzylbromid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC303056-10G
PC303056-5G
PC303056-1G
APOSPC303056-10GEA
0
EUR
APOSPC303056-10G
APOSPC303056-5G
APOSPC303056-1G
3-Chlor-4-(trifluormethyl)benzylbromid 98%
3-Chlor-4-(trifluormethyl)benzylbromid
Formel:
C₈H₅BrClF₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11519327 CAS-Nummer: 361393-92-2 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...