3-Chlor-5-(trifluormethoxy)benzonitril

Lieferant: Apollo Scientific
Gefahr

PC303060-10G PC303060-5G PC303060-1G
APOSPC303060-10GEA 474 EUR
APOSPC303060-10G APOSPC303060-5G APOSPC303060-1G
3-Chlor-5-(trifluormethoxy)benzonitril
3-Chlor-5-(trifluormethoxy)benzonitril
Formel: C₈H₃ClF₃NO
Molecular Weight: 221,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660309
CAS-Nummer: 886503-29-3
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR