3-Ethoxy-2,4-difluorphenol 98%
Lieferant: Apollo Scientific
PC303064-10G
PC303064-5G
PC303064-25G
APOSPC303064-10GEA
405
EUR
APOSPC303064-10G
APOSPC303064-5G
APOSPC303064-25G
3-Ethoxy-2,4-difluorphenol 98%
3-Ethoxy-2,4-difluorphenol
Formel:
C₈H₈F₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258727 CAS-Nummer: 1017778-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...