3-Methoxy-4-(trifluormethyl)benzylalkohol 98%

Lieferant: Apollo Scientific

PC303070-10G PC303070-5G PC303070-25G
APOSPC303070-10GEA 459 EUR
APOSPC303070-10G APOSPC303070-5G APOSPC303070-25G
3-Methoxy-4-(trifluormethyl)benzylalkohol 98%
3-Methoxy-4-(trifluormethyl)benzylalkohol
Formel: C₉H₉F₃O₂
Molecular Weight: 206,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18398590
CAS-Nummer: 276861-64-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR