2,3-Difluor-4-ethoxybenzonitril 97%

Lieferant: Apollo Scientific

Synonyme: 4-Ethoxy-2,3-difluorobenzonitrile

PC303082-10G PC303082-5G PC303082-1G
APOSPC303082-10GEA 42.2 EUR
APOSPC303082-10G APOSPC303082-5G APOSPC303082-1G
2,3-Difluor-4-ethoxybenzonitril 97%
2,3-Difluor-4-ethoxybenzonitril
Formel: C₉H₇F₂NO
Molecular Weight: 183.1548 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04973770
CAS-Nummer: 126162-96-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR