4'-Fluor-3',5'-dimethylacetophenon 95%

Lieferant: Apollo Scientific

PC303087-10G PC303087-5G PC303087-1G
APOSPC303087-10GEA 705 EUR
APOSPC303087-10G APOSPC303087-5G APOSPC303087-1G
4'-Fluor-3',5'-dimethylacetophenon 95%
4'-Fluor-3',5'-dimethylacetophenon
Formel: C₁₀H₁₁FO
Molecular Weight: 166.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660269
CAS-Nummer: 42444-20-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR