4-Fluoro-3-methyl-5-(trifluoromethyl)benzyl bromide 98%

Lieferant: Apollo Scientific

Synonyme: 4-Fluoro-3-methyl-5-(trifluoromethyl)benzyl bromide

PC303092-10G PC303092-5G PC303092-1G
APOSPC303092-10GEA 746 EUR
APOSPC303092-10G APOSPC303092-5G APOSPC303092-1G
4-Fluoro-3-methyl-5-(trifluoromethyl)benzyl bromide 98%
4-Fluoro-3-methyl-5-(trifluoromethyl)benzyl bromide
Formel: C₉H₇BrF₄
Molecular Weight: 271,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201072
CAS-Nummer: 1373921-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR