6-Chloro-2,3-difluorobenzyl bromide 97%
Lieferant: Apollo Scientific
Synonyme:
6-Chloro-2,3-difluorobenzyl bromide
PC303099-10G
PC303099-5G
PC303099-1G
APOSPC303099-10GEA
351
EUR
APOSPC303099-10G
APOSPC303099-5G
APOSPC303099-1G
6-Chloro-2,3-difluorobenzyl bromide 97%
6-Chloro-2,3-difluorobenzyl bromide
Formel:
C₇H₄BrClF₂ Molecular Weight: 241,46 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22201040 CAS-Nummer: 1373921-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...