6-Chlor-2-fluor-3-methoxyphenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC303101-10G
PC303101-5G
PC303101-25G
APOSPC303101-10GEA
420
EUR
APOSPC303101-10G
APOSPC303101-5G
APOSPC303101-25G
6-Chlor-2-fluor-3-methoxyphenol 98%
6-Chlor-2-fluor-3-methoxyphenol
Formel:
C₇H₆ClFO₂ Molecular Weight: 176,57 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832270 CAS-Nummer: 1017777-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...