4-(4-Chlorphenoxy)-3-(trifluormethyl)anilin 98%

Lieferant: Apollo Scientific

PC303108-10G PC303108-5G PC303108-1G
APOSPC303108-10GEA 405 EUR
APOSPC303108-10G APOSPC303108-5G APOSPC303108-1G
4-(4-Chlorphenoxy)-3-(trifluormethyl)anilin 98%
4-(4-Chlorphenoxy)-3-(trifluormethyl)anilin
Formel: C₁₃H₉ClF₃NO
Molecular Weight: 287,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01908501
CAS-Nummer: 57688-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR