(2-Chlor-4-(trifluormethyl)phenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Chloro-4-(trifluoromethyl)phenyl)acetic acid

PC303152-1G PC303152-5G PC303152-25G
APOSPC303152-1GEA 66.7 EUR
APOSPC303152-1G APOSPC303152-5G APOSPC303152-25G
(2-Chlor-4-(trifluormethyl)phenyl)essigsäure 98%
(2-Chlor-4-(trifluormethyl)phenyl)essigsäure
Formel: C₉H₆ClF₃O₂
Molecular Weight: 238,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194288
CAS-Nummer: 601513-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR