2,4-Difluor-5-methylbenzaldehyd 98%
Lieferant: Apollo Scientific
Synonyme:
4,6-Difluor-m-methylbenzaldehyd, 2,4-Difluor-5-tolualdehyd, 2,4-Difluor-5-tolylaldehyd, 4,6-Difluor-m-tolylaldehyd, 4,6-Difluor-m-tolualdehyd
PC303301-5G
PC303301-10G
PC303301-1G
APOSPC303301-5GEA
277
EUR
APOSPC303301-5G
APOSPC303301-10G
APOSPC303301-1G
2,4-Difluor-5-methylbenzaldehyd 98%
2,4-Difluoro-5-methylbenzaldehyde
Formel:
C₈H₆F₂O Molecular Weight: 156.13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12196967 CAS-Nummer: 315204-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...