1-Chloro-1,3,3,3-tetrafluoropropene
Lieferant: Apollo Scientific
PC3034-1G
PC3034-5G
PC3034-25G
APOSPC3034-1GEA
0
EUR
APOSPC3034-1G
APOSPC3034-5G
APOSPC3034-25G
1-Chloro-1,3,3,3-tetrafluoropropene
1-Chloro-1,3,3,3-tetrafluoropropene
Formel:
C₃HClF₄ Siedepunkt: 19…20 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038011 CAS-Nummer: 460-71-9 UN: 3161 ADR: 2.1, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...