6-(2,2,2-Trifluoroethoxy)pyridine-3-thio-carboxamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-(2,2,2-Trifluoroethoxy)thionicotinamide
PC3048-1G
PC3048-250MG
APOSPC3048-1GEA
348
EUR
APOSPC3048-1G
APOSPC3048-250MG
6-(2,2,2-Trifluoroethoxy)pyridine-3-thio-carboxamide
6-(2,2,2-Trifluoroethoxy)pyridine-3-thio-carboxamide
Formel:
C₈H₇F₃N₂OS Molecular Weight: 236,22 g/mol Schmelzpunkt: 120…123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00277205 CAS-Nummer: 175277-59-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...