(5-Fluoropyridin-2-yl)methanol

Lieferant: Apollo Scientific

Synonyme: 5-Fluoro-2-(hydroxymethyl)pyridine

PC305009-10G PC305009-1G PC305009-5G
APOSPC305009-10GEA 400 EUR
APOSPC305009-10G APOSPC305009-1G APOSPC305009-5G
(5-Fluoropyridin-2-yl)methanol
(5-Fluoropyridin-2-yl)methanol
Formel: C₆H₆FNO
Molecular Weight: 127,1200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027049
CAS-Nummer: 802325-29-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR