2,5-Bis(2,2,2-trifluoroethoxy)benzenesulphonylchloride 97%

Lieferant: Apollo Scientific
Gefahr

PC3053M-1G PC3053M-5G
APOSPC3053M-1GEA 52.2 EUR
APOSPC3053M-1G APOSPC3053M-5G
2,5-Bis(2,2,2-trifluoroethoxy)benzenesulphonylchloride 97%
2,5-Bis(2,2,2-trifluoroethoxy)benzenesulphonylchloride
Formel: C₁₀H₇ClF₆O₄S
Molecular Weight: 372.67 g/mol
Schmelzpunkt: 48…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052841
CAS-Nummer: 152457-95-9
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR