5-Fluor-m-kresol 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-Fluor-3-hydroxy-5-methylbenzol, 3-Fluor-5-methylphenol, 3-Fluor-5-hydroxytoluol

PC3069-5G PC3069-10G PC3069-1G
APOSPC3069-5GEA 49.3 EUR
APOSPC3069-5G APOSPC3069-10G APOSPC3069-1G
5-Fluor-m-kresol 97%
5-Fluor-m-kresol
Formel: C₇H₇FO
Molecular Weight: 126.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115885
CAS-Nummer: 216976-31-7
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR