3,4'-Bis(trifluormethyl)benzhydrol 97%

Lieferant: Apollo Scientific

PC3086E-1G PC3086E-5G
APOSPC3086E-1GEA 84.1 EUR
APOSPC3086E-1G APOSPC3086E-5G
3,4'-Bis(trifluormethyl)benzhydrol 97%
3,4'-Bis(trifluormethyl)benzhydrol
Formel: C₁₅H₁₀F₆O
Molecular Weight: 320,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00045091
CAS-Nummer: 203915-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR