2,2'-Bis(trifluoromethyl)benzidine 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,2'-Bis(trifluoromethyl)-[1,1'-biphenyl]-4,4'-diamine, 4,4'-Diamino-2,2'-bis(trifluoromethyl)biphenyl
PC3086K-1G
PC3086K-5G
PC3086K-25G
APOSPC3086K-1GEA
14.7
EUR
APOSPC3086K-1G
APOSPC3086K-5G
APOSPC3086K-25G
2,2'-Bis(trifluoromethyl)benzidine 97%
2,2'-Bis(trifluoromethyl)benzidine
Formel:
C₁₄H₁₀F₆N₂ Molecular Weight: 320.24 g/mol Schmelzpunkt: 180…181 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190155 CAS-Nummer: 341-58-2 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...