2,4-Bis(trifluormethyl)benzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC3087-25G
PC3087-5G
PC3087-1G
APOSPC3087-25GEA
133
EUR
APOSPC3087-25G
APOSPC3087-5G
APOSPC3087-1G
2,4-Bis(trifluormethyl)benzoesäure 98%
2,4-Bis(trifluormethyl)benzoesäure
Formel:
C₉H₄F₆O₂ Molecular Weight: 258,12 g/mol Schmelzpunkt: 108…111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042496 CAS-Nummer: 32890-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...